siRNA Details
| Gene Symbol: ABO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020469.2 | NP_065202.2 | 7 | 1148 | ||
| GenBank mRNA | AK302203.1 | - | 6 | 996 | ||
| JX519569.1 | - | 7 | 1123 | |||
| JX519570.1 | - | 7 | 1123 | |||
| U15197.1 | - | 1 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABO| Gene Symbol: ABO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020469.2 | NP_065202.2 | 7 | 1148 | ||
| GenBank mRNA | AK302203.1 | - | 6 | 996 | ||
| JX519569.1 | - | 7 | 1123 | |||
| JX519570.1 | - | 7 | 1123 | |||
| U15197.1 | - | 1 | 74 | |||