siRNA Details
| Gene Symbol: Zfp397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027007.2 | NP_081283.2 | 2, 3 | 694 | ||
| GenBank mRNA | AK013086.1 | - | 2, 3 | 657 | ||
| AK032001.1 | - | 2, 3 | 697 | |||
| AY338468.1 | - | 1, 2 | 518 | |||
| BC021456.1 | - | 2, 3 | 678 | |||

Zfp397
Genomic Map