siRNA Details
| Gene Symbol: Zfp397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027007.2 | NP_081283.2 | 4 | 1495 | ||
| GenBank mRNA | AK032001.1 | - | 4 | 1498 | ||
| AY338468.1 | - | 3 | 1319 | |||
| BC021456.1 | - | 4 | 1479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp397| Gene Symbol: Zfp397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027007.2 | NP_081283.2 | 4 | 1495 | ||
| GenBank mRNA | AK032001.1 | - | 4 | 1498 | ||
| AY338468.1 | - | 3 | 1319 | |||
| BC021456.1 | - | 4 | 1479 | |||