siRNA Details
| Gene Symbol: Rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027009.2 | NP_081285.1 | 4 | 372 | ||
| GenBank mRNA | AK013095.1 | - | 4 | 360 | ||
| AK042393.1 | - | 4 | 373 | |||
| AK145126.1 | - | 4 | 399 | |||
| AK165891.1 | - | 4 | 367 | |||
| AK167047.1 | - | 4 | 373 | |||
| BC026795.1 | - | 4 | 343 | 4015134 | ||

Rfc3
Genomic Map