siRNA Details
| Gene Symbol: Cst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027024.3 | NP_081300.1 | 3 | 502 | ||
| GenBank mRNA | AK005665.1 | - | 3 | 480 | ||
| AY600990.2 | - | 3 | 520 | |||
| BC048681.1 | - | 3 | 502 | 6774445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst13| Gene Symbol: Cst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027024.3 | NP_081300.1 | 3 | 502 | ||
| GenBank mRNA | AK005665.1 | - | 3 | 480 | ||
| AY600990.2 | - | 3 | 520 | |||
| BC048681.1 | - | 3 | 502 | 6774445 | ||