siRNA Details
| Gene Symbol: Izumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027034.1 | NP_081310.1 | 4, 5 | 658 | ||
| GenBank mRNA | AK005867.1 | - | 4, 5 | 206 | ||
| BC099579.1 | - | 4, 5 | 657 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Izumo3| Gene Symbol: Izumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027034.1 | NP_081310.1 | 4, 5 | 658 | ||
| GenBank mRNA | AK005867.1 | - | 4, 5 | 206 | ||
| BC099579.1 | - | 4, 5 | 657 | |||