siRNA Details
| Gene Symbol: 1700008P02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027048.1 | NP_081324.1 | 2 | 472 | ||
| GenBank mRNA | AK005786.1 | - | 2 | 474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700008P02Rik| Gene Symbol: 1700008P02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027048.1 | NP_081324.1 | 2 | 472 | ||
| GenBank mRNA | AK005786.1 | - | 2 | 474 | ||