siRNA Details
| Gene Symbol: Slc38a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027052.3 | NP_081328.2 | 14 | 1502 | ||
| GenBank mRNA | AB055004.1 | - | 14 | 1478 | ||
| AK005887.1 | - | 1 | 29 | |||
| AK051023.1 | - | 14 | 1503 | |||
| AK164292.1 | - | 14 | 1445 | |||
| AK167539.1 | - | 14 | 1484 | |||
| AK169349.1 | - | 13 | 1491 | |||
| AY027919.1 | - | 14 | 1541 | |||
| BC024072.1 | - | 14 | 1427 | 5097293 | ||
| BC024123.1 | - | 14 | 1459 | 5098067 | ||
| BC031717.1 | - | 14 | 1420 | 4195569 | ||

Slc38a4
Genomic Map