siRNA Details
| Gene Symbol: MICB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289160.1 | NP_001276089.1 | 5 | 1037 | ||
| NM_001289161.1 | NP_001276090.1 | 5 | 1007 | |||
| NM_005931.4 | NP_005922.2 | 5 | 1136 | |||
| GenBank mRNA | AF021221.1 | - | 4 | 922 | ||
| AF021222.1 | - | 4 | 922 | |||
| AF021223.1 | - | 4 | 922 | |||
| AF021224.1 | - | 4 | 922 | |||
| AF021225.1 | - | 4 | 922 | |||
| AF021226.1 | - | 4 | 922 | |||
| AK291927.1 | - | 5 | 1134 | |||
| AK300791.1 | - | 5 | 1037 | |||
| AK303655.1 | - | 4 | 1271 | |||
| AK314228.1 | - | 5 | 1041 | |||
| AY885251.1 | - | 6 | 1088 | |||
| BC044218.1 | - | 5 | 971 | 6025253 | ||
| U95729.1 | - | 4 | 923 | |||
| U95730.1 | - | 4 | 923 | |||
| U95731.1 | - | 4 | 923 | |||
| U95732.1 | - | 4 | 923 | |||
| U95733.1 | - | 4 | 923 | |||
| U95734.1 | - | 4 | 923 | |||
| X91625.1 | - | 5 | 997 | |||

MICB
Genomic Map