siRNA Details
| Gene Symbol: 1700013G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027063.2 | NP_081339.1 | 2 | 814 | ||
| GenBank mRNA | AK005952.1 | - | 2 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700013G24Rik| Gene Symbol: 1700013G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027063.2 | NP_081339.1 | 2 | 814 | ||
| GenBank mRNA | AK005952.1 | - | 2 | 813 | ||