siRNA Details
| Gene Symbol: 1700016C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027077.2 | NP_081353.1 | 2, 3 | 227 | ||
| GenBank mRNA | AK006014.1 | - | 2, 3 | 228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700016C15Rik| Gene Symbol: 1700016C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027077.2 | NP_081353.1 | 2, 3 | 227 | ||
| GenBank mRNA | AK006014.1 | - | 2, 3 | 228 | ||