siRNA Details
| Gene Symbol: 1700016C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027077.2 | NP_081353.1 | 4 | 422 | ||
| GenBank mRNA | AK006014.1 | - | 4 | 423 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 1700016C15Rik
1700016C15Rik| Gene Symbol: 1700016C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027077.2 | NP_081353.1 | 4 | 422 | ||
| GenBank mRNA | AK006014.1 | - | 4 | 423 | ||
 Back To Top
 Back To Top