siRNA Details
| Gene Symbol: 2300005B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081961.1 | NP_001075430.1 | 2, 3 | 175 | ||
| GenBank mRNA | AB272582.1 | - | 2, 3 | 163 | ||
| AK009031.1 | - | 1, 2 | 98 | |||
| BC115610.1 | - | 2, 3 | 175 | 40102714 | ||
| BC115611.1 | - | 2, 3 | 175 | 40102716 | ||

2300005B03Rik
Genomic Map