siRNA Details
| Gene Symbol: 1700029J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033148.3 | NP_001028320.1 | 5 | 2254 | ||
| GenBank mRNA | AK006501.1 | - | 6 | 848 | ||
| AK156041.1 | - | 5 | 2256 | |||
| BC107256.2 | - | 6 | 737 | 40052445 | ||

1700029J07Rik
Genomic Map