siRNA Details
| Gene Symbol: Tsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164578.1 | NP_001158050.1 | 5 | 561 | ||
| NM_175146.4 | NP_780355.2 | 5 | 552 | |||
| GenBank mRNA | AK044504.1 | - | 5 | 528 | ||
| BC025008.1 | - | 5 | 515 | |||
| BC037444.1 | - | 4 | 416 | |||
| BC049668.1 | - | 5 | 534 | 6535058 | ||
| BG083382.2 | - | 4 | 425 | |||
| CF732210.1 | - | 5 | 525 | |||

Tsr2
Genomic Map