siRNA Details
| Gene Symbol: Tsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175146.4 | NP_780355.2 | 4, 5 | 480 | ||
| GenBank mRNA | AK044504.1 | - | 4, 5 | 456 | ||
| BC025008.1 | - | 4, 5 | 443 | |||
| BC037444.1 | - | 3, 4 | 344 | |||
| BC049668.1 | - | 4, 5 | 462 | 6535058 | ||
| BG083382.2 | - | 3, 4 | 353 | |||

Tsr2
Genomic Map