siRNA Details
| Gene Symbol: 1700030J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027103.2 | NP_081379.1 | 3 | 1027 | ||
| GenBank mRNA | AK006553.1 | - | 3 | 1032 | ||
| AK030075.1 | - | 3 | 1028 | |||
| AK031529.1 | - | 3 | 912 | |||
| BC049630.1 | - | 3 | 956 | |||
| BC100501.1 | - | 3 | 1047 | 30917058 | ||
| BC137750.1 | - | 3 | 965 | 8860769 | ||
| BC137751.1 | - | 3 | 965 | 8860770 | ||

1700030J22Rik
Genomic Map