siRNA Details
| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027107.1 | NP_081383.1 | 4 | 608 | ||
| GenBank mRNA | AK009156.1 | - | 4 | 575 | ||
| AK146257.1 | - | 4 | 598 | |||
| BC027639.1 | - | 4 | 553 | 1069092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ten1| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027107.1 | NP_081383.1 | 4 | 608 | ||
| GenBank mRNA | AK009156.1 | - | 4 | 575 | ||
| AK146257.1 | - | 4 | 598 | |||
| BC027639.1 | - | 4 | 553 | 1069092 | ||