siRNA Details
| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027107.1 | NP_081383.1 | 3 | 321 | ||
| GenBank mRNA | AK009156.1 | - | 3 | 288 | ||
| AK146257.1 | - | 3 | 311 | |||
| AK153854.1 | - | 2 | 910 | |||
| BC027639.1 | - | 3 | 266 | 1069092 | ||

Ten1
Genomic Map