siRNA Details
| Gene Symbol: Trnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081156.2 | NP_001074625.1 | 2 | 1576 | ||
| GenBank mRNA | AY142705.1 | - | 2 | 990 | ||
| BC049168.1 | - | 2 | 1532 | 6491759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trnp1| Gene Symbol: Trnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081156.2 | NP_001074625.1 | 2 | 1576 | ||
| GenBank mRNA | AY142705.1 | - | 2 | 990 | ||
| BC049168.1 | - | 2 | 1532 | 6491759 | ||