siRNA Details
| Gene Symbol: Wdr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027113.2 | NP_081389.1 | 1 | 839 | ||
| GenBank mRNA | AK009247.1 | - | 1 | 754 | ||
| AK149554.1 | - | 810 | ||||
| AK165100.1 | - | 703 | ||||
| BC064045.1 | - | 1 | 839 | 1245342 | ||
| EF011616.1 | - | 1 | 837 | |||

Wdr5b
Genomic Map