siRNA Details
| Gene Symbol: Nkpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027116.1 | NP_081392.1 | 4 | 1259 | ||
| GenBank mRNA | BC118925.2 | - | 4 | 1259 | 40088590 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkpd1| Gene Symbol: Nkpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027116.1 | NP_081392.1 | 4 | 1259 | ||
| GenBank mRNA | BC118925.2 | - | 4 | 1259 | 40088590 | |