siRNA Details
| Gene Symbol: Bst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198095.2 | NP_932763.1 | 1 | 143 | ||
| GenBank mRNA | AK154399.1 | - | 1 | 136 | ||
| BC027328.1 | - | 1 | 177 | 4009434 | ||
| BC056638.1 | - | 1 | 150 | 5065707 | ||
| BC087949.1 | - | 1 | 135 | 6768735 | ||

Bst2
Genomic Map