siRNA Details
| Gene Symbol: Fam65c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080708.2 | NP_001074177.1 | 2, 3 | 193 | ||
| GenBank mRNA | AK158217.1 | - | 2, 3 | 219 | ||
| BC130272.1 | - | 2, 3 | 193 | 40143210 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam65c| Gene Symbol: Fam65c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080708.2 | NP_001074177.1 | 2, 3 | 193 | ||
| GenBank mRNA | AK158217.1 | - | 2, 3 | 219 | ||
| BC130272.1 | - | 2, 3 | 193 | 40143210 | ||