siRNA Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027122.3 | NP_081398.2 | 2 | 861 | ||
| GenBank mRNA | AK089539.1 | - | 2 | 862 | ||
| AK217168.1 | - | 2 | 289 | |||
| BC019171.1 | - | 2 | 888 | 5003354 | ||
| BC085305.1 | - | 2 | 925 | 6399056 | ||

Mfsd3
Genomic Map