siRNA Details
| Gene Symbol: Otop3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027132.2 | NP_081408.2 | 2 | 424 | ||
| GenBank mRNA | AB087505.1 | - | 2 | 379 | ||
| AK043266.1 | - | 2 | 379 | |||
| BC145613.1 | - | 2 | 341 | 9054179 | ||
| BC150803.1 | - | 2 | 341 | 9087714 | ||

Otop3
Genomic Map