siRNA Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110209.1 | NP_001103679.1 | 5 | 466 | ||
| NM_027133.3 | NP_081409.1 | 5 | 466 | |||
| GenBank mRNA | AK009311.1 | - | 5 | 440 | ||
| AK014897.1 | - | 5 | 364 | |||
| AK053593.1 | - | 5 | 435 | |||
| AK136506.1 | - | 5 | 441 | |||
| AK137445.1 | - | 5 | 467 | |||
| AK143497.1 | - | 5 | 435 | |||
| AK173241.1 | - | 5 | 398 | |||
| AY312281.1 | - | 5 | 387 | |||
| BC057961.1 | - | 5 | 393 | 5353629 | ||
| BC060153.1 | - | 4 | 305 | |||

Lnp
Genomic Map