siRNA Details
| Gene Symbol: Txndc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175150.3 | NP_780359.2 | 3 | 687 | ||
| GenBank mRNA | AK009878.1 | - | 3 | 615 | ||
| AK079517.1 | - | 3 | 634 | |||
| AK160051.1 | - | 3 | 656 | |||
| AK167701.1 | - | 3 | 595 | |||
| AK167838.1 | - | 3 | 653 | |||
| AK167977.1 | - | 3 | 652 | |||
| BC029015.1 | - | 2 | 423 | |||
| BC062180.1 | - | 3 | 687 | 6336176 | ||
| BC094275.1 | - | 3 | 523 | |||

Txndc15
Genomic Map