siRNA Details
| Gene Symbol: Cdk3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009918.1 | - | 5 | 985 | ||
| AK166387.1 | - | 2 | 472 | |||
| BC116895.1 | - | 5 | 895 | |||
| BC119034.1 | - | 5 | 895 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk3-ps| Gene Symbol: Cdk3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009918.1 | - | 5 | 985 | ||
| AK166387.1 | - | 2 | 472 | |||
| BC116895.1 | - | 5 | 895 | |||
| BC119034.1 | - | 5 | 895 | |||