siRNA Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197991.2 | NP_932108.2 | 5 | 661 | ||
| GenBank mRNA | AK150499.1 | - | 5 | 566 | ||
| AK160274.1 | - | 5 | 578 | |||
| AK169010.1 | - | 5 | 612 | |||
| AK171657.1 | - | 5 | 575 | |||
| AK189635.1 | - | 1 | 13 | |||
| AY761096.1 | - | 5 | 554 | |||
| AY761098.1 | - | 5 | 554 | |||
| BC006065.1 | - | 3 | 298 | |||
| BC020179.1 | - | 5 | 556 | 4913500 | ||

Emc10
Genomic Map