siRNA Details
| Gene Symbol: Tatdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175151.4 | NP_780360.1 | 6 | 393 | ||
| GenBank mRNA | AK010232.1 | - | 5 | 478 | ||
| AK042248.1 | - | 6 | 394 | |||
| AK161972.1 | - | 6 | 478 | |||
| BC034895.1 | - | 4 | 266 | |||
| BC061187.1 | - | 6 | 390 | 30277245 | ||

Tatdn1
Genomic Map