siRNA Details
| Gene Symbol: Camsap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163749.1 | NP_001157221.1 | 17 | 3991 | ||
| NM_027171.3 | NP_081447.2 | 17 | 3988 | |||
| GenBank mRNA | AK009960.1 | - | 5 | 575 | ||
| AK081728.1 | - | 17 | 3763 | |||
| AK220401.1 | - | 11 | 4195 | |||
| BC037466.1 | - | 7 | 1826 | |||
| BC043033.1 | - | 9 | 2606 | |||
| BC043081.1 | - | 17 | 3647 | |||
| BC048787.1 | - | 17 | 3636 | 6489361 | ||
| KC426930.1 | - | 19 | 3710 | |||
| KC426931.1 | - | 19 | 3707 | |||
| KC426932.1 | - | 18 | 3677 | |||
| KC426933.1 | - | 18 | 3674 | |||
| KC426934.1 | - | 18 | 3662 | |||
| KC426935.1 | - | 18 | 3659 | |||
| KC426936.1 | - | 17 | 3629 | |||
| KC426937.1 | - | 17 | 3626 | |||
| KC426938.1 | - | 19 | 2462 | |||
| KC426939.1 | - | 19 | 2459 | |||
| KC426940.1 | - | 18 | 2429 | |||
| KC426941.1 | - | 18 | 2426 | |||
| KC426942.1 | - | 18 | 2414 | |||
| KC426943.1 | - | 18 | 2411 | |||
| KC426944.1 | - | 17 | 2381 | |||
| KC426945.1 | - | 17 | 2378 | |||

Camsap3
Genomic Map