siRNA Details
| Gene Symbol: Camsap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163749.1 | NP_001157221.1 | 11 | 2744 | ||
| NM_027171.3 | NP_081447.2 | 11 | 2741 | |||
| GenBank mRNA | AK081728.1 | - | 11 | 2516 | ||
| AK220401.1 | - | 7 | 1886 | |||
| BC037466.1 | - | 1 | 579 | |||
| BC043033.1 | - | 3 | 1359 | |||
| BC043081.1 | - | 11 | 2400 | |||
| BC048787.1 | - | 11 | 2389 | 6489361 | ||
| KC426930.1 | - | 13 | 2463 | |||
| KC426931.1 | - | 13 | 2460 | |||
| KC426932.1 | - | 12 | 2430 | |||
| KC426933.1 | - | 12 | 2427 | |||
| KC426934.1 | - | 12 | 2415 | |||
| KC426935.1 | - | 12 | 2412 | |||
| KC426936.1 | - | 11 | 2382 | |||
| KC426937.1 | - | 11 | 2379 | |||

Camsap3
Genomic Map