siRNA Details
| Gene Symbol: Camsap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163749.1 | NP_001157221.1 | 17 | 4035 | ||
| NM_027171.3 | NP_081447.2 | 17 | 4032 | |||
| GenBank mRNA | AK009960.1 | - | 5 | 619 | ||
| AK081728.1 | - | 17 | 3807 | |||
| AK220401.1 | - | 11 | 4239 | |||
| BC037466.1 | - | 7 | 1870 | |||
| BC043033.1 | - | 9 | 2650 | |||
| BC043081.1 | - | 17 | 3691 | |||
| BC048787.1 | - | 17 | 3680 | 6489361 | ||
| KC426930.1 | - | 19 | 3754 | |||
| KC426931.1 | - | 19 | 3751 | |||
| KC426932.1 | - | 18 | 3721 | |||
| KC426933.1 | - | 18 | 3718 | |||
| KC426934.1 | - | 18 | 3706 | |||
| KC426935.1 | - | 18 | 3703 | |||
| KC426936.1 | - | 17 | 3673 | |||
| KC426937.1 | - | 17 | 3670 | |||
| KC426938.1 | - | 19 | 2506 | |||
| KC426939.1 | - | 19 | 2503 | |||
| KC426940.1 | - | 18 | 2473 | |||
| KC426941.1 | - | 18 | 2470 | |||
| KC426942.1 | - | 18 | 2458 | |||
| KC426943.1 | - | 18 | 2455 | |||
| KC426944.1 | - | 17 | 2425 | |||
| KC426945.1 | - | 17 | 2422 | |||

Camsap3
Genomic Map