siRNA Details
| Gene Symbol: Slc52a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164819.1 | NP_001158291.1 | 2 | 670 | ||
| NM_001164820.1 | NP_001158292.1 | 2 | 670 | |||
| NM_027172.3 | NP_081448.2 | 2 | 738 | |||
| GenBank mRNA | AK009850.1 | - | 2 | 621 | ||
| AK029091.1 | - | 2 | 715 | |||
| AK047618.1 | - | 2 | 671 | |||
| AK154970.1 | - | 2 | 720 | |||
| AK170158.1 | - | 2 | 699 | |||
| BC016127.1 | - | 2 | 639 | 4910227 | ||
More Information
Set Membership: |
|

Slc52a3
Genomic Map