siRNA Details
Gene Symbol: 2310079G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027173.1 | NP_081449.1 | 1 | 497 | ||
GenBank mRNA | AK010217.1 | - | 1 | 499 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310079G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027173.1 | NP_081449.1 | 1 | 497 | ||
GenBank mRNA | AK010217.1 | - | 1 | 499 |