siRNA Details
| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081406.1 | NP_001074875.1 | 1, 2 | 243 | ||
| GenBank mRNA | AK010419.1 | - | 1, 2 | 236 | ||
| AK144741.1 | - | 1, 2 | 244 | |||
| BC022648.1 | - | 1, 2 | 197 | |||
| BC075711.1 | - | 1, 2 | 225 | |||
| BC117053.1 | - | 1, 2 | 222 | |||
| BC117055.1 | - | 1, 2 | 222 | |||

Lrr1
Genomic Map