siRNA Details
| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178378.3 | NP_848465.1 | 11 | 1294 | ||
| GenBank mRNA | AK010269.1 | - | 8 | 843 | ||
| AK146422.1 | - | 9 | 1032 | |||
| BC049990.1 | - | 11 | 1278 | 6310844 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcg| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178378.3 | NP_848465.1 | 11 | 1294 | ||
| GenBank mRNA | AK010269.1 | - | 8 | 843 | ||
| AK146422.1 | - | 9 | 1032 | |||
| BC049990.1 | - | 11 | 1278 | 6310844 | ||