siRNA Details
| Gene Symbol: MLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040109.1 | NP_001035198.1 | 3 | 293 | ||
| NM_001184698.1 | NP_001171627.1 | 3 | 293 | |||
| NM_002418.2 | NP_002409.1 | 3 | 293 | |||
| GenBank mRNA | BC069675.1 | - | 3 | 288 | 7262661 | |
| BC112314.1 | - | 3 | 257 | 8327782 | ||
| BC143981.1 | - | 3 | 257 | 9052499 | ||
| Y00695.1 | - | 3 | 277 | |||

MLN
Genomic Map