siRNA Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027184.1 | NP_081460.1 | 6 | 856 | ||
| GenBank mRNA | AK036084.1 | - | 6 | 624 | ||
| AK036978.1 | - | 6 | 816 | |||
| AK172246.1 | - | 6 | 948 | |||
| BC052463.1 | - | 6 | 856 | 6834748 | ||

Ipmk
Genomic Map