siRNA Details
| Gene Symbol: Rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027186.2 | NP_081462.1 | 6 | 828 | ||
| GenBank mRNA | AK010289.1 | - | 6 | 557 | ||
| BC047996.1 | - | 6 | 815 | 6509910 | ||
| BY576068.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpain| Gene Symbol: Rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027186.2 | NP_081462.1 | 6 | 828 | ||
| GenBank mRNA | AK010289.1 | - | 6 | 557 | ||
| BC047996.1 | - | 6 | 815 | 6509910 | ||
| BY576068.1 | - | 1 | 38 | |||