siRNA Details
| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177561.3 | NP_808229.1 | 4 | 496 | ||
| GenBank mRNA | AK029604.1 | - | 3 | 348 | ||
| AK145298.1 | - | 4 | 516 | |||
| BC030888.1 | - | 2 | 263 | |||
| BC039916.1 | - | 4 | 498 | 4950034 | ||

Usp46
Genomic Map