siRNA Details
| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177561.3 | NP_808229.1 | 8 | 1078 | ||
| GenBank mRNA | AK029604.1 | - | 7 | 930 | ||
| AK145298.1 | - | 8 | 1098 | |||
| BC030888.1 | - | 6 | 845 | |||
| BC039916.1 | - | 8 | 1080 | 4950034 | ||

Usp46
Genomic Map