siRNA Details
| Gene Symbol: Gemin7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027189.2 | NP_081465.1 | 2 | 360 | ||
| GenBank mRNA | AK010303.1 | - | 2 | 362 | ||
| BC028527.1 | - | 2 | 356 | 1362579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gemin7| Gene Symbol: Gemin7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027189.2 | NP_081465.1 | 2 | 360 | ||
| GenBank mRNA | AK010303.1 | - | 2 | 362 | ||
| BC028527.1 | - | 2 | 356 | 1362579 | ||