siRNA Details
| Gene Symbol: Nup37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027191.2 | NP_081467.2 | 3 | 325 | ||
| NM_028334.4 | NP_082610.1 | 3 | 343 | |||
| GenBank mRNA | AK010370.1 | - | 3 | 269 | ||
| AK012890.1 | - | 3 | 339 | |||
| AK040463.1 | - | 3 | 326 | |||
| AK165537.1 | - | 3 | 289 | |||
| BC011102.1 | - | 3 | 309 | 3982076 | ||
| BC032180.1 | - | 2 | 253 | 5151847 | ||
| CJ072834.1 | - | 3 | 345 | |||

Nup37
Genomic Map