siRNA Details
| Gene Symbol: Casz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159344.1 | NP_001152816.1 | 5 | 628 | ||
| NM_027195.2 | NP_081471.2 | 5 | 628 | |||
| GenBank mRNA | BC075673.1 | - | 5 | 630 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casz1| Gene Symbol: Casz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159344.1 | NP_001152816.1 | 5 | 628 | ||
| NM_027195.2 | NP_081471.2 | 5 | 628 | |||
| GenBank mRNA | BC075673.1 | - | 5 | 630 | ||