siRNA Details
| Gene Symbol: Zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027198.1 | NP_081474.1 | 5 | 438 | ||
| GenBank mRNA | AA027411.1 | - | 1 | 30 | ||
| AK010472.1 | - | 5 | 418 | |||
| BC109343.2 | - | 5 | 396 | 40053371 | ||
| BQ554850.1 | - | 5 | 441 | |||

Zswim7
Genomic Map