siRNA Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153195.2 | NP_694875.2 | 2, 3 | 519 | ||
| GenBank mRNA | AK147797.1 | - | 2, 3 | 471 | ||
| AK147907.1 | - | 2, 3 | 438 | |||
| AK150190.1 | - | 2, 3 | 294 | |||
| AK150626.1 | - | 2, 3 | 477 | |||
| AK151025.1 | - | 2, 3 | 477 | |||
| AK152513.1 | - | 2, 3 | 477 | |||
| AK182909.1 | - | 1, 2 | 131 | |||
| BC032153.1 | - | 2, 3 | 461 | 5054087 | ||

Fbxo7
Genomic Map