siRNA Details
| Gene Symbol: Leng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027203.3 | NP_081479.1 | 2 | 542 | ||
| GenBank mRNA | AK005095.1 | - | 2 | 173 | ||
| AK037634.1 | - | 2 | 189 | |||
| BC023108.1 | - | 2 | 542 | 4021762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Leng1| Gene Symbol: Leng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027203.3 | NP_081479.1 | 2 | 542 | ||
| GenBank mRNA | AK005095.1 | - | 2 | 173 | ||
| AK037634.1 | - | 2 | 189 | |||
| BC023108.1 | - | 2 | 542 | 4021762 | ||