siRNA Details
| Gene Symbol: 1600015I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081273.2 | NP_001074742.2 | 1 | 1734 | ||
| GenBank mRNA | BC085280.1 | - | 1 | 1735 | ||
| BC099440.1 | - | 1 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600015I10Rik| Gene Symbol: 1600015I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081273.2 | NP_001074742.2 | 1 | 1734 | ||
| GenBank mRNA | BC085280.1 | - | 1 | 1735 | ||
| BC099440.1 | - | 1 | 371 | |||